SPILFYTER® Mercsorb® Mercury Amalgamation Powder, NPS
Catalog #: 15551-838
Supplier: NPS
Specifications
- Description:Mercury Amalgamation Powder
- Size:500 g
- Cat. no.:15551-838
- Supplier No.:520500
Specifications
About this item
This Mercury Amalgamation Powder is used to convert elemental mercury into an amalgam, which stops dangerous mercury vapors from being emitted.
Customers Who Bought This Also Bought