Orders LinkContactUsLinkComponent
pCMV-Script Vectors, Agilent Technologies
pCMV-Script Vectors, Agilent Technologies
pCMV-Script Vectors, Agilent Technologies
Supplier:  
Certificates
pCMV-Script Vectors, Agilent Technologies
pCMV-Script Vectors, Agilent Technologies
0 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
pCMV-Script PCR cloning system is a polymerase chain reaction (PCR) cloning method that can be performed in 1-hour without adding bases to the primers. The kit permits the efficient cloning of PCR fragments with a high yield and a low rate of false positives.
  • CMV promoter for high-level protein expression in mammalian cells
  • Large multiple cloning site contains 15 unique sites
  • Clone PCR products generated with any PCR enzyme
Caution: For Research Use Only. Not for Use in Diagnostic Procedures.
Product Family Options