Order Entry
Canada
ContactUsLinkComponent
 
Absolutely Total RNA Purification Kits, Agilent Technologies
 
undefined
Absolutely Total RNA Purification Kits, Agilent Technologies
 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Micro
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Absolutely RNA Nanoprep Kit

 

Frequently Bought Together