Micro Fluorescence Cuvette with PTFE Cover, Type 507, FireflySci
Supplier: FIREFLYSCI, INC.
Certificates
About this item
The Type 507 cuvettes are made with Optical Glass, UV Quartz, IR Quartz with transmission values and it includes PTFE covers
- Ensuring high quality optical polishing with no defects
- Highly accurate results only need to test sample once
- Excellent transmission values
- Four windows and base optically polished
Fire-fused to ensure that cuvettes are robust and durable, optically polished windows and base.
Customers Who Bought This Also Bought