Short Sleeve Denim Work Shirts, Dickies®
Supplier: Dickies
Certificates
About this item
Dickies® style WS300.
- Box pleat back
- Replacement buttons on front placket
- Button-down collar
- 8 oz. denim, 100% cotton, enzyme washed
Specifications
- Catalog No:
- 76162-774
- 76162-776
- 76162-778
- 76162-780
- 76162-782
- 76162-784
- 76162-786
- 76162-788
- 76162-790
- 76162-792
- Model:
- WS300
- WS300
- WS300
- WS300
- WS300
- WS300
- WS300
- WS300
- WS300
- WS300
- Garment application:
- Industrial, General Purpose
- Industrial, General Purpose
- Industrial, General Purpose
- Industrial, General Purpose
- Industrial, General Purpose
- Industrial, General Purpose
- Industrial, General Purpose
- Industrial, General Purpose
- Industrial, General Purpose
- Industrial, General Purpose
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- Weight:
- 8 oz.
- 8 oz.
- 8 oz.
- 8 oz.
- 8 oz.
- 8 oz.
- 8 oz.
- 8 oz.
- 8 oz.
- 8 oz.
Specifications
Customers Who Bought This Also Bought