Versaflo™ High Efficiency Filter, 3M™
Catalog # 10152-834
Supplier: SCS
Specifications
- Description:High Efficiency Filter
- Cat. no.:10152-834
- Supplier No.:00051131373582
Specifications
About this item
TR-6710N is a High Efficiency (HE) particulate filter for use with TR-600 Series PAPRs. The filter attaches directly to the motor/blower. 3M™ recommends the use of the TR-6700FC filter cover to help protect the filter from physical damage. The TR-6700FC filter cover is also used to hold the TR-6600 prefilter or TR-662 spark arrestor in place.
- High efficiency filter
- Bulk packaging for convenience
- Simple to install