Specifications
- Description:Arduino Leonardo Development Board
- Cat. no.:470237-742
Specifications
About this item
Based on the ATmega32u4.
- Used for sensing and instrumentation
- Perfect for advanced programming use
- ATmega32u4 8bit AVR RISC based microcontroller
- Arduino Leonardo can be powered via the micro USB connection or with an external power supply
- The board can operate on an external supply of 6 to 20 V
- Built in LED connected to digital pin 13; When the pin is high value, the LED is on
- The board appears as a generic keyboard and mouse, and can be prog to control these input devices
- USB Over current protection
This board has 20 digital input/output pins (of which 7 can be used as PWM outputs and 12 as analog inputs), a 16MHz crystal oscillator, a micro USB connection, a power jack, an ICSP header, and a reset button. It contains everything needed to support the microcontroller, simply connect it to a computer with a USB cable or power it with a AC to DC adapter or battery to get started. The board differs from all preceding boards in that the ATmega32u4 has built in USB communication, eliminating the need for a secondary processor. This allows the Leonardo to appear to a connected computer as a mouse and keyboard, in addition to a virtual (CDC) serial / COM port. It also has other implications for the behaviour of the board. The maximum length and width of the Leonardo PCB are 2.7" and 2.1".