If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Enzyme type:Recombinant
- Species:Human
- Size:10 μg
- Enzyme name:Ube2C Cg01
- Cat. no.:102498-876
- Supplier No.:CG01-10UG
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
- Consistent sizing options
- Datasheet included
- High purity, low endotoxin levels
Recombinant human ubiquitin-conjugating enzyme E2 C is produced by our E.coli expression system and the target gene encoding Met1-Pro179 is expressed with a 6His tag at the N-terminus.