It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
- Consistent sizing options
- Datasheet included
- High purity, low endotoxin levels
Recombinant human LILRA3 is produced by our mammalian expression system and the target gene encoding Thr19-Glu439 is expressed with a 6His tag at the C-terminus.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Enzyme type:Recombinant
- Species:Human
- Enzyme name:Ilt6 Cc83