Order Entry
Canada
ContactUsLinkComponent
Recombinant Gal-beta-1
Recombinant Gal-beta-1
Certificates
Recombinant Gal-beta-1

Specifications

  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    6-sialyltransferase II|GalNAc alpha-2|ST6GalNAc II|6-sialyltransferase|3-GalNAc alpha-2|Sialyltransferase 7B|SIAT7-B
  • Protein/peptide name:
    Gal-beta-1
  • Species:
    Mouse

Specifications

Frequently Bought Together