Human Recombinant Coag Factor Xiii A Chain (C-6His)
Supplier: Bon Opus Biosciences
Certificates
About this item
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
- Consistent Sizing options
- Datasheet Included
- High Purity, Low Endotoxin levels
Recombinant Human Coagulation Factor XIII A Chain is produced by our Mammalian expression system and the target gene encoding Gly39-Met732 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Specifications
- Enzyme type:Recombinant
- Species:Human
- Enzyme name:Coag Factor Xiii A Chain (C-6His)
Specifications
Frequently Bought Together