Recombinant Butyrophilin subfamily 3 member A1
Supplier: Bon Opus Biosciences
Certificates
About this item
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
- Consistent Sizing options
- Datasheet Included
- High Purity, Low Endotoxin levels
Recombinant Human Butyrophilin Subfamily 3 Member A1 is produced by our Mammalian expression system and the target gene encoding Gln30-Gly254 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Specifications
- Protein/peptide type:Recombinant
- Protein/peptide name:Butyrophilin subfamily 3 member A1
- Species:Human
Specifications
Frequently Bought Together