SP Wilmad-LabGlass Bend Adapters with Hose Connection and Standard Taper Joint, SP Industries
Catalog # 14206-312
Supplier: SP Industries
Specifications
- ST Joint:24/40
- Cat. no.:14206-312
- Hose Connection O.D.:⅜"
- Supplier No.:LG-1702-102
Specifications
About this item
These borosilicate glass adapters have a 90° bend to hose connector at top and a standard taper inner joint at bottom.