ZR Small-RNA™ PAGE Recovery Kit, Zymo Research
Catalog # 76020-408
Supplier: Zymo Research
CAS Number:
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Small RNA (miRNA, siRNA, shRNA)
- Sample type:Gel matrix
- Target molecule:RNA
- Description:ZR small-RNA™ PAGE Recovery Kit
- Size:20 Preps
- Description:ZR small-RNA™ PAGE Recovery Kit
- Yield:10 µg RNA (binding capacity), ≥6 µl (elution volume)
- Cat. no.:76020-408
- Format:Spin column
Specifications
About this item
The ZR small-RNA™ PAGE Recovery Kit provides an easy and efficient method for the extraction of high quality small RNAs from polyacrylamide gels (native and/or denatured).
- For efficient recovery of small RNA fragments from polyacrylamide gels
- Zymo-Spin™ column technology allows RNA to be eluted into minimal volumes
- Compatible with up to 25% (w/v) polyacrylamide
- RNA is ready for all subsequent analysis and molecular manipulation
The ZR small-RNA™ PAGE Recovery Kit is a refinement of the "crush and soak" method that incorporates a unique buffer system together with Zymo-Spin™ column technologies for improved recovery and added convenience. The recovered RNA can be concentrated into volumes as small as 6 µL, making it ideal for many downstream enzymatic reactions and manipulations.