Extraction Reagents 2 (5X), (10 ml), Enzo Life Sciences
CA95043-696
:
- Description:Extraction reagent 2 (5X)
- Size:10 mL
- Formulation:Liquid. TRIS buffer containing detergents.
- Cat. no.:CA95043-696
- Supplier No.:80-1502
Concentrated lysis buffer for preparation of cell and tissue extracts.
TRIS buffer containing detergents.