ContactUsLinkComponent
Photosynthesis and Cellular Respiration Model Set
Photosynthesis and Cellular Respiration Model Set
Catalog # 470232-064
Supplier:  Mega Molecules
undefined
Photosynthesis and Cellular Respiration Model Set
Catalog # 470232-064
Supplier:  Mega Molecules
Supplier Number:  BC84
Specifications
  • Description:
    Photosynthesis and Respiration Model Set
  • Cat. no.:
    470232-064
Specifications
About this item
Hands on demonstration of the formation of ATP.
  • Perfect for middle school students
Photosynthesis and cellular respiration are the fundamental processes in the flow of energy and the cycling of matter. Students are given the task of building models of water and carbon dioxide molecules, which are converted into glucose and oxygen molecules at the giant chloroplast. The glucose and oxygen molecules then move to the mitochondria where the students reassemble them into carbon dioxide and water molecules, generating ATP.