Molecular Geometry Model Set (VSEPR)
Molecular Geometry (VSEPR)
Catalog # 470232-056
Supplier: Mega Molecules
: MG67
Jump to:
Product Details & Documents
Easily illustrate the 5 VSEPR models.
- Perfect for high school and college students
Five universal VSEPR models allows the students to build linear, planar triangular, tetrahedral, trigonal bipyramidal, and octahedral structures. When the student builds the molecules over a white sheet of paper and uses the white links to represent the lone pairs of electrons on the central atom, the molecular geometries can be easily visualized.
Specifications
- Description:Molecular Geometry (VSEPR)