Organic Chemistry Molecular Model Set
Catalog # 470232-050
Supplier: Mega Molecules
Specifications
- Description:Organic Chemistry Molecular Model Set
- Cat. no.:470232-050
Specifications
About this item
Easily convert between boat and chair conformations with this organic molecular model set.
- Perfect size to carry in a backpack
- 140 pieces
- Plastic case and instructions included
- 1 year FREE replacement guarantee against atom breakage
The set contains 140 durable plastic components that can be used to build thousands of molecules. The carbon-carbon bonds are easy to rotate, allowing boat to chair conformational changes in seconds. The flexible links allow you to create single, double, and triple bonds. The snap-closure prevents spills and the compartments keep the atom models and bonds conveniently separated. The model building instructions include pictures that show how to assemble the atom models.