Annexin V Binding Buffer, BioLegend®
Supplier: Biolegend
Certificates
About this item
Annexin V Binding Buffer has been formulated for flow cytometric labeling of apoptotic cells with Annexin V reagents. The buffer is provided as a ready-to-use solution.
Caution: Store between 2 and 8 °C.
Specifications
- Storage temperature:2 – 8 °C