Oakton® PCTS Testr™ 50 Waterproof Pocket pH/Cond/TDS/Salinity Tester, Premium 50 Series, Cole-Parmer
Catalog # 76208-092
Supplier: Antyila Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Calibration:Up to 3 points
- Temperature range:0 – 60 °C (32 – 140 °F)
- Description:Waterproof Pocket pH/Cond/TDS/Salinity Tester
- Environmentally Preferable:
- Temperature accuracy:0 – 50 °C (±0.5 °C/±0.9 °F); 50 – 60 °C (±1.0 °C/±1.8 °F)
- Temperature resolution:0.1 °C (0.1 °F)
- Battery life:150 hrs.
- Automatic buffer recognition:USA 4.01, 7.00, 10.01 and NIST 4.01, 6.86, 9.18
- Conductivity accuracy:±1% full scale
- Conductivity range:0 µS - 20 mS
- Conductivity resolution:0.1 μS, 1 μS, 0.01 mS
- Salinity accuracy:±1% full scale
- Salinity range:0.00 – 10.0 ppt
- Salinity resolution:0.01 ppt
- TDS accuracy:±1% full scale
- TDS range:0 ppm – 10.0 ppt
- TDS resolution:10 ppt
- Temperature compensation:ATC
- pH accuracy:±0.01 pH
- pH range:–1 to 15
- pH resolution:0.01 pH
- Battery type:Four AAA 1.5 V
- Cat. no.:76208-092
Specifications
About this item
Ultimate tester for pH, conductivity, TDS, salinity, and temperature!
- Combine measurement capabilities of three testers in one convenient pocket-sized unit
- Up to 3 point pH calibration with NIST or USA auto buffer recognition
- Conductivity, TDS, and salinity feature auto ranging based on the concentration of your sample
- Waterproof
- LCD display
Redesigned leakproof cap functions as a sample/calibration cup, hydrating cup, and stabilizing base to stand the meter upright.
Delivery: Includes protective plastic storage case, lanyard, and 4 AAA batteries.