SP Wilmad-LabGlass Reaction Kettle Heating Mantles, SP Industries
Supplier: SP Industries
Certificates
About this item
These all-glass constructed heating mantles are for use with LG-8071 through LG-8089 reaction kettles. They provide even heat distribution.
- Heating circuits have two short lead-in wires connected by a Hubbell plug to a 4' long, 2-wire type SJT cord
- Depth given is the interior measurement
- Wattage requirements are for 115 V circuits
Customers Who Bought This Also Bought