Hercules® 3041 Gloves, HexArmor
Supplier: HexArmor
Certificates
About this item
The Hercules® 3041 has a full coverage design and pre-curved shape with a specialized silicon gripping surface on the palm, and the highest levels of needle and cut protection available on the market.
- Highest needlestick-resistance available
- Specialized silicon gripping surface on palm
- Launderable
The Hercules® NSR offers the highest levels of cut-resistance (ANSI/ISEA A9, CE Level 5) and full-hand needlestick protection by layering HexArmor’s exclusively-licensed SuperFabric® brand material. Needlestick protection verified using 25 gauge needles in the ASTM 1342-05 test.
Specifications
- Catalog No:
- 76208-028
- 76208-024
- 76208-026
- 76208-466
- 76208-468
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- EN norm:
- En388 4522
- En388 4522
- En388 4522
- En388 4522
- En388 4522