Helix® 2066 Gloves, HexArmor
Supplier: HexArmor
Certificates
About this item
The lightweight, German-engineered Phynomic® Series 2066 gloves feature an ANSI/ISEA cut level A3 protective Dyneema® Diamond shell that radiates heat away from workers’ hands so they stay comfortable, cool, and dry all day. Coated in Germany and silicone-free, the flexible foam nitrile palm coating provides superior abrasion resistance and grip in dry, wet, or oily situations.
- Knit: 15-gauge Dyneema® Diamond shell provides lightweight, cool comfort
- Coating: Flexible foam nitrile palm coating provides superior grip and abrasion resistance
- Knit wrist helps prevent dirt and debris from entering the glove
The Dyneema® Diamond shell keeps hands cool and comfortable while the flexible foam nitrile palm coating provides excellent abrasion resistance and grip in dry, wet, or oily situations.
Certifications: Oeko-Tex and ProDerm certified. REACH Compliant for being void of harsh chemicals. Helix® 2066 gloves provide an ANSI/ISEA A3 cut level.
Specifications
- Catalog No:
- 76207-978
- 76207-980
- 76207-974
- 76207-976
- 76207-984
- 76207-982
- 76207-972
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Gauge:
- 15-Gauge
- 15-Gauge
- 15-Gauge
- 15-Gauge
- 15-Gauge
- 15-Gauge
- 15-Gauge
- EN norm:
- En388 4X42C
- En388 4X42C
- En388 4X42C
- En388 4X42C
- En388 4X42C
- En388 4X42C
- En388 4X42C
Specifications
Related Information
Helix 2066
Datasheet
Hexarmor EMR Protection Solutions HexArmor Linecard HexArmor PBI Matrix
Product Literature
Frequently Bought Together