Econo™ Metzenbaum-Lahey Dissecting Scissors, Floor Grade, Sklar®
Catalog # 76192-452
Supplier: Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Length:4.5"
- Material:Stainless steel
- Environmentally Preferable:
- Curvature:Curved
- Cat. no.:76192-452
- Blade Type:Blunt, Smooth
Specifications
About this item
Econo™ Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
Econo™ Metzenbaum-Lahey Dissecting Scissors are not recommended for cutting sutures, drains, or heavy tissue. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.