ContactUsLinkComponent
Brunner Ligature Needle, OR Grade, Sklar®
Brunner Ligature Needle, OR Grade, Sklar®
Catalog # 76193-006
Supplier:  Sklar
Brunner Ligature Needle, OR Grade, Sklar®
Catalog # 76193-006
Supplier:  Sklar
Supplier Number:  26-1615
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Sterility:
    Non sterile
  • Length:
    9.5"
  • Material:
    Stainless steel
  • Curvature:
    Curved
  • Cat. no.:
    76193-006
  • Tip Style:
    Blunt, Fenestrated
  • Handle:
    Hollow
Specifications
About this item
Reusable, curved needle made from stainless steel. Premium OR-Grade.
  • Reusable
  • Hollow handle
  • Latex-free