If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Length:4"
- Material:Stainless steel
- Tip Configuration:Smooth
- Curvature:Straight
- Cat. no.:76192-488
Specifications
About this item
Econo™ Littauer Cutting Forceps are a non-ratcheted, plier handle instrument used primarily for cutting nail and bone.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
As nail nippers they are used for nail debridement. They may also be used to nip delicate bones found in the facial and nasal region, or small animal (mice) veterinary procedures. The short, sharp blades are able to produce enough force to cut through thick nail and small bone, but delicate enough not to cause trauma to surrounding tissue or organs. This is particularly helpful with nail debridement which should be completed in short, small sections to avoid damage to the hyponychium. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.