Moynihan Forceps are ratcheted, hemostatic forceps used for grasping sections of tissue.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
Sklar® Moynihan Artery Forceps are ratcheted, hemostatic forceps typically used for grasping large sections of tissue or passing sutures in abdominal procedures. They have long shanks and short jaws with full, horizontal serrations. The tips are blunt and may be used for dissection.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.