Orders LinkContactUsLinkComponent
Laboratory Frames and Supports
Laboratory Frames and Supports
Supplier:   OHAUS CORP.
Laboratory Frames and Supports
Supplier:  OHAUS CORP.
Certificates
Laboratory Frames and Supports
Laboratory Frames and Supports
2 Options Available Below
Product Details & Documents
Provides secure support for holding glassware at elevated positions.
  • Stainless steel construction
  • Two sizes available to meet your laboratory needs
Small lab-frame equipped for vertical mounting. Great for glassware set-ups in small laboratories or where space is limited. Frame measures 24 x 24" (610 x 610 mm). Medium lab-frame can be mounted either veristically or horizontally, which is convenient for distillation or general set-up. Frame measures 24 x 48" (610 x 1219 mm). Bases are both 18" (457 mm) wide for stability. Base may be permanently mounted to bench top with screws (included).
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 470235-412
    Small Lab Frame
    Small Lab Frame
    24 x 24"
    Log in for availability
    Log in for availability
    Each
    Each - $1,888.90
    $1,888.90
    Each
       $1,888.90
    Supplier Number: 30392323
  • 470235-414
    Medium Lab Frame
    Medium Lab Frame
    24 x 48"
    Log in for availability
    Log in for availability
    Each
    Each - $2,653.10
    $2,653.10
    Each
       $2,653.10
    Supplier Number: 30392324