Orders LinkContactUsLinkComponent
Lambda Systems, Agilent Technologies
Lambda Systems, Agilent Technologies
Lambda Systems, Agilent Technologies
Supplier:  
Certificates
Lambda Systems, Agilent Technologies
Lambda Systems, Agilent Technologies
0 Options Available Below
Product Details & Documents
Lambda DASH II and Lambda FIX II vectors offer traditional genomic cloning of 9 to 23 kB fragments.
These vectors are used for genomic library construction contain T3 and T7 RNA promoters for rapid gene walking and high-resolution gene mapping.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options