Human Recombinant NAP-2 (from E. coli)
Catalog # 77660-754
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:25 µg
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P02775
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Platelet basic protein|MDGF|LDGF|PBP|CXCL7, Neutrophil activating peptide 2|PPBP
- Protein/peptide name:NAP-2
- Purity:≥95%
- Molecular weight:7.6 kDa
- Sequence:AELRCMCIKT TSGIHPKNIQ SLEVIGKGTH CNQVEVIATL KDGRKICLDP DAPRIKKIVQ KKLAGDESAD
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Cat. no.:77660-754
Specifications
About this item
Neutrophil activating peptide 2 (NAP-2), also known as CXCL7, is a member of the CXC family of chemokines. NAP-2 is a carboxyl-terminal fragment produced by proteolytic cleavage of the platelet basic protein (PBP). NAP-2 is released from platelets and binds to the receptors CXCR1 and CXCR2 to chemoattract and activate neutrophils during inflammatory events.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.