Order Entry
Canada
Orders LinkContactUsLinkComponent
Mini Solvent Pump, Burkle

Mini Solvent Pump, Burkle

Mini Solvent Pump, 1.8 L/min. (gpm)
Catalog # 76203-678
Supplier:   BURKLE INC
Supplier #:  5603-4000
Mini Solvent Pump, Burkle
Catalog # 76203-678
Supplier:  BURKLE INC
Mini Solvent Pump, Burkle
Mini Solvent Pump, Burkle

Mini Solvent Pump, 1.8 L/min. (gpm)

Price
$2,189.63
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The Mini solvent pump is a high-quality hand pump for small containers such as bottles, canisters and tanks up to about 10 liters (2.64 gallons).

  • For reliable filling and dosing of solvents and readily flammable liquids
  • Ideal for 1 to 5 l (0.26 to 1.32 gal.) containers
  • Connecting thread GL 45
  • Parts in contact with medium made of PTFE and stainless steel AISI 304 (1.4301), AISI 316L (1.4404)
  • PTFE hose

Simple principle, highly effective–The hand pump works according to the excess pressure principle. An excess pressure is built up in the container with the pump ball, forcing the liquid outward. In this way only the hose, pumping tube and discharge come in contact with the liquid being filled. To ensure safety, a pressure relief valve protects the solvent pump against excessive pressure build-up, for example due to sunshine.

The Mini solvent pump is therefore especially well suited for use with high-purity liquids and readily flammable media such as acetone, isopropanol, ethanol and methanol, etc.

Filling in hazardous zones–The grounding cable included with delivery can easily be connected to the solvent pump with a bunch plug, thus diverting electrostatic charges so that flammable media can be filled safely. Flexible applications–The solvent pump allows for filling from a wide range of container sizes. The flexible PTFE hose that conveys liquid out of the container can be shortened to any length. This ensures ideal residual draining, even from bottles and canisters with a vaulted bottom.

Precise removal of liquids–The self-closing discharge (with auto-stop function) of the Mini solvent pump ensures absolutely precise filling with no dripping at all. Dosing is even easy in very small containers such as test tubes, which present no problems. A hose can optionally be fastened onto the discharge spout (internal diameter 6 mm (0.24")) thus making it possible to fill the container from a greater distance.

The Mini solvent pump is especially compact and very well balanced, which makes it possible to fill without tipping, even from very small containers. Thanks to its sturdy design and high-quality materials, the Mini solvent pump consistently ensures neat and reliable filling throughout its long service life, easily and conveniently.

Accessories information: Adapter available as accessory for S 40, GL 38 and GL 32.

Delivery information: Including grounding cable 2 m (6.56 ft.).

Documents

Specifications
  • Capacity:
    1.8 L/min. (gpm)
  • Description:
    Mini Solvent Pump
  • Cat. No.:
    76203-678
  • Hose length:
    6 x 8 mm (0.24 x 0.31"); 0.4 m (1.31')
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Thread Adapter, PTFE, GL 45 - GL 32
    Thread Adapter, PTFE, GL 45 - GL 32
    76203-592
    Log in for availability
    Each - 166,87 $

Product Information

  • Catalog #
    Availability
    Price
  • Thread Adapter, PTFE, GL 45 - S 40
    Thread Adapter, PTFE, GL 45 - S 40
    76203-680
    Log in for availability
    Each - 168,34 $

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.