MsGM™ Mesangial Cell Growth Medium
MsGM™ Mesangial Cell Growth Medium; is formulated for the growth of renal mesangial cells. MsGM™ contains 5% serum.
: Kit includes MsBM basal medium 500 mL and MsGM SingleQuot kit with supplement and growth factors,1×Red cap vial with GA-1000 0.50 ml, 1×bottle FBS 25.00 ml.
Frequently Bought Together