YeaStar™ RNA Kit, Zymo Research
Catalog # 77002-008
Supplier: Zymo Research
CAS Number:
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Yeast/Fungal RNA
- Sample type:Yeast/Fungi
- Target molecule:RNA
- Description:YeaStar™ RNA Kit
- Size:40 Preps
- Description:YeaStar™ RNA Kit
- Yield:≤ 25 µg of total RNA
- Cat. no.:77002-008
Specifications
About this item
The YeaStar™ RNA Kit provides all the necessary reagents for RNA isolation from a broad spectrum of fungi including: Aspergillus fumigatus, Aspergillus nidulans, Aspergillus nivens var. aureus, Candida albicans, Pichia pastoris, Saccharomyces cerevisiae, Schizosaccharomyces pombe.
- Recovery of purified RNA from a wide range of fungus species using Zymo-Spin™ column technology
- Omits the use of glass beads and organic denaturants
- Eluted RNA is suitable for use in RT-PCR or other RNA-based procedures
The kit can be used for the purification of high-quality, total RNA from any fungus that can be lysed by yeast lytic enzyme. The kit facilitates the purification of 10 to 25 µg of total RNA from 1 to 1.5 mL of cultured cells.
Delivery information: Kit includes YR digestion buffer, YR lysis buffer, RNA wash buffer1 (concentrate), DNase/RNase-free water, Zymolyase, Zymolyase storage buffer, Zymo-Spin™ IIIC columns, collection tubes and instruction manual.