VWR® ESD Micro Denier Scrub Pant
Supplier: VWR International
Certificates
About this item
This style of Scrub Pant is widely used in pharmaceutical cleanroom operations, and meets/exceeds the recommendations outlined in the IEST-RP-CC-003.4, Garment Considerations for Cleanrooms and Other Controlled Environments.
- Combination elastic back and drawstring front for secure comfort
- Leg opening with elastic back and three-position snaps for closure
- Double-needle felled outer seams for extra durability
- Garments include color-coded loops for easy size identification
- All edges are pre-serged or laser cut for lint limitation
These scrubs provide an excellent base-layer of protection for any cleanroom garment system. Made with Sandwash material, this lightweight micro-denier fabric is woven from 100% continuous-filament polyester to eliminate any source for lint.
Specifications
- Catalog No:
- 76202-686
- 76202-688
- 76202-690
- 76202-700
- 76202-680
- 76202-682
- 76202-684
- 76202-692
- 76202-698
- Garment application:
- Cleanroom, Pharmaceutical
- Cleanroom, Pharmaceutical
- Cleanroom, Pharmaceutical
- Cleanroom, Pharmaceutical
- Cleanroom, Pharmaceutical
- Cleanroom, Pharmaceutical
- Cleanroom, Pharmaceutical
- Cleanroom, Pharmaceutical
- Cleanroom, Pharmaceutical
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 100% continuous-filament polyester
- 100% continuous-filament polyester
- 100% continuous-filament polyester
- 100% continuous-filament polyester
- 100% continuous-filament polyester
- 100% continuous-filament polyester
- 100% continuous-filament polyester
- 100% continuous-filament polyester
- 100% continuous-filament polyester