ContactUsLinkComponent
Catabolic Buffer, Xenotech
Catabolic Buffer, Xenotech
Catalog # 103307-306
Supplier:  Xenotech LLC
CAS Number:  
Catabolic Buffer, Xenotech
Catalog # 103307-306
Supplier:  Xenotech LLC
Supplier Number:  K5200
CAS Number:  
Specifications
  • Description:
    10x Catabolic Buffer
  • Size:
    1 mL
  • Cat. no.:
    103307-306
  • Supplier No.:
    K5200
Specifications
About this item
Catabolic Buffer has been formulated and optimized to extract the most in vitro catabolic performance from isolated human lysosomes and rat tritosomes. Although DTT is present in the buffer, it can easily be removed.