DuPont™ Tychem® 2000 SFR Protective Coveralls
Supplier: DuPont
Certificates
About this item
Tychem® 2000 SFR Protective Coveralls provide chemical and secondary flame protection in a lightweight garment.
- Respirator-fit hood lined with ProShield® 6 SFR fabric
- Covered, braided elastic at hood
- Nylon zipper with large metal pull
- Chin flap with double-sided adhesive tape
- Storm flap covers zipper, which can be sealed by the wearer with double-sided adhesive strip to prevent intrusion at zipper
- Covered, braided elastic openings at wrist and ankle for tighter fit
- Taped seams
Tychem® 2000 SFR garments are intended to be worn over primary flame resistant garments. In the event that a flash fire occurs, they do not ignite and do not contribute to additional burn injury if appropriate flame-resistant personal protective equipment, such as DuPont™ Nomex®, is worn beneath.
Certifications: Thickness meets ASTM D1777.