Order Entry
Canada
ContactUsLinkComponent
 
Anti-FCGR3A Mouse Monoclonal Antibody (CF568) [clone: ICO-116]
 :  Biotium
 
Anti-FCGR3A Mouse Monoclonal Antibody (CF568) [clone: ICO-116]
 :  Biotium

 

  • Catalog No:
  • 75888-614
  • 75888-616
  • Antigen symbol:
  • FCGR3A
  • FCGR3A
  • Antigen name:
  • CD16
  • CD16
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • ICO-116
  • ICO-116
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 372679
  • Unigene ID: 372679
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

 

Frequently Bought Together