About this item
This antibody recognizes a common epitope of human major histocompatibility (MHC) class II antigens, HLA-DR and DP. Human MHC class II antigens are transmembrane glycoproteins composed of an chain (36 kDa) and a chain (27 kDa). They are expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, and thymic epithelial cells and are also present on activated T lymphocytes. Human MHC class II genes are located in the HLA-D region that encodes at least six and ten chain genes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They, therefore, have a critical role in the initiation of the immune response. It has been shown that some autoimmune diseases are associated with certain class II alleles.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Type: Primary
Antigen: HLA-DPB1 & HLA-DRA
Clonality: Monoclonal
Clone: Clone Bra-14
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75960-932
- 75960-934
- Antigen symbol:
- HLA-DPB1 & HLA-DRA
- HLA-DPB1 & HLA-DRA
- Antigen name:
- MHC II
- MHC II
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- Bra-14
- Bra-14
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 347270 (HLA-DP) , 520048 (HLA-DR)
- Unigene ID: 347270 (HLA-DP) , 520048 (HLA-DR)
- Isotype:
- IgG3 kappa
- IgG3 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide