About this item
This antibody recognizes a protein of 140 kDa, which is identified as the complement receptor 2 (CR2)/CD21. Its epitope is located in 5-8 short consensus repeats (SCRs). This MAb is highly specific to CR2 and shows no cross-reaction with CR1. This protein is expressed strongly on mature B cells, follicular dendritic cells and weakly on immature thymocytes and T lymphocytes. In B-cell ontogeny, CD21 appears after the pre-B-stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells. CD21 expression is also gradually lost after stimulation of B cells in vitro. CD21 functions as receptor for C3d, C3dg and iC3b Complement components, for EBV and for IFNalpha. CD21 binds to CD23 and associates with CD19, CD81 and Leu13 to form a large signal-transduction complex involved in B cell activation. MAb FR5A10 can be used for EBV receptor studies, interactions between B and T cells especially through CD23, human complement receptor (CR2) studies and IFN-alpha receptor studies.
Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: Clone FR5A10
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75890-890
- 75890-892
- Antigen symbol:
- CR2
- CR2
- Antigen name:
- CD21
- CD21
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- FR5A10
- FR5A10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 445757
- Unigene ID: 445757
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide