Order Entry
Canada
ContactUsLinkComponent
 
Anti-CR2 Mouse Monoclonal Antibody (CF594) [clone: FR5A10]
 :  Biotium
 
Anti-CR2 Mouse Monoclonal Antibody (CF594) [clone: FR5A10]
 :  Biotium

 

  • Catalog No:
  • 75890-920
  • 75890-918
  • Antigen symbol:
  • CR2
  • CR2
  • Antigen name:
  • CD21
  • CD21
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • FR5A10
  • FR5A10
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 445757
  • Unigene ID: 445757
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

 

Frequently Bought Together