Order Entry
Canada
ContactUsLinkComponent
Anti-MYOD1 Mouse Monoclonal Antibody (CF405S) [clone: 5.8A MYD712]
Anti-MYOD1 Mouse Monoclonal Antibody (CF405S) [clone: 5.8A MYD712]
Supplier:  Biotium
Certificates
Anti-MYOD1 Mouse Monoclonal Antibody (CF405S) [clone: 5.8A MYD712]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75964-756
  • 75964-758
  • Antigen symbol:
  • MYOD1
  • MYOD1
  • Antigen name:
  • MyoD1
  • MyoD1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • 5.8A MYD712
  • 5.8A MYD712
  • Reactivity:
  • Chicken, Human, Rat, Mouse
  • Chicken, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 181768
  • Unigene ID: 181768
  • Isotype:
  • IgGs
  • IgGs
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together