CD2 interacts through its amino-terminal domain with the extracellular domain of CD58 (also designated CD2 ligand) to mediate cell adhesion. CD2/CD58 binding can enhance antigen-specific T cell activation. CD2 is a transmembrane glycoprotein that is expressed on peripheral blood T lymphocytes, NK cells and thymocytes. CD58 is a heavily glycosylated protein with a broad tissue distribution in hematopoietic and other cells, including endothelium. Interaction between CD2 and its counter receptor LFA3 (CD58) on opposing cells optimizes immune system recognition, thereby facilitating communication between helper T lymphocytes and antigen-presenting cells, as well as between cytolytic effectors and target cells.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: Clone LFA2/600
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Non-human primates, Pig
- Catalog No:
- 75890-282
- 75890-280
- Antigen symbol:
- CD2
- CD2
- Antigen name:
- CD2
- CD2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- LFA2/600
- LFA2/600
- Reactivity:
- Pig, Non-human primates, Human
- Pig, Non-human primates, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 523500
- Unigene ID: 523500
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Concentration:
- 0.2 mg/mL
- 0.2 mg/mL
- Storage buffer:
- PBS/0.05% BSA/0.05% azide
- PBS/0.05% BSA/0.05% azide