Anti-MUC1 Mouse Monoclonal Antibody [clone: MUC1/520]
75963-750
: Biotium
- Antibody type:Primary
- Antigen name:Mucin 1 / EMA / Episialin / CD227
- Antigen symbol:MUC1
- Clonality:Monoclonal
- Clone:MUC1/520
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:50 μL
- Gene ID:Unigene ID: 89603
- Storage buffer:PBS, no BSA, no azide
- Concentration:1 mg/mL
- Cat. no.:75963-750
- Supplier No.:BNUM0520-50
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinomas. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver.
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: Clone MUC1/520
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human