Order Entry
Canada
ContactUsLinkComponent
Anti-CD2 Mouse Monoclonal Antibody (CF405S) [clone: 1E7E8.G4]
Anti-CD2 Mouse Monoclonal Antibody (CF405S) [clone: 1E7E8.G4]
Supplier:  Biotium
Certificates
Anti-CD2 Mouse Monoclonal Antibody (CF405S) [clone: 1E7E8.G4]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75889-896
  • 75889-894
  • Antigen symbol:
  • CD2
  • CD2
  • Antigen name:
  • CD2
  • CD2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • 1E7E8.G4
  • 1E7E8.G4
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 523500
  • Unigene ID: 523500
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together