Order Entry
Canada
ContactUsLinkComponent
Anti-PGF Mouse Monoclonal Antibody (CF488A) [clone: PLGF93]
Anti-PGF Mouse Monoclonal Antibody (CF488A) [clone: PLGF93]
Supplier:  Biotium
Certificates
Anti-PGF Mouse Monoclonal Antibody (CF488A) [clone: PLGF93]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75969-920
  • 75969-922
  • Antigen symbol:
  • PGF
  • PGF
  • Antigen name:
  • PLGF
  • PLGF
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • PLGF93
  • PLGF93
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 252820
  • Unigene ID: 252820
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together