Order Entry
Canada
ContactUsLinkComponent
Anti-KRT19 Mouse Monoclonal Antibody (CF568) [clone: A53-B/A2.26]
Anti-KRT19 Mouse Monoclonal Antibody (CF568) [clone: A53-B/A2.26]
Supplier:  Biotium
Certificates
Anti-KRT19 Mouse Monoclonal Antibody (CF568) [clone: A53-B/A2.26]
Supplier:  Biotium

Specifications

  • Catalog No:
  • 75919-590
  • 75919-588
  • Antigen symbol:
  • KRT19
  • KRT19
  • Antigen name:
  • Cytokeratin 19
  • Cytokeratin 19
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • A53-B/A2.26
  • A53-B/A2.26
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 654568
  • Unigene ID: 654568
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

Specifications

Frequently Bought Together