Orders LinkContactUsLinkComponent
Reagent Troughs
Supplier:   TWD Scientific
Reagent Troughs
Supplier:  TWD Scientific
Certificates
Reagent Troughs
2 Options Available Below
Product Details & Documents
Reagent Troughs are made of polypropylene. Troughs support a disposable polypropylene trough liner. Liners are made of thin, disposable, lightweight polypropylene.
  • V-shaped bottom and angled sides for maximum reagent recovery
  • Usable with up to 12-channel pipettors
  • Designed for SBS format applications and compatible with robotic systems
  • Reusable
Perfect for 12 or 8-channel pipettes and compatible eith robotic systems.
Product Family Options
Product Information
  • Material
    Capacity
    Availability
    Price
  • 76197-170
    Reagent Trough
    Reagent Trough
    PP
    100 ml
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $385.15
    $385.15
    Case of 24
       $385.15
    Supplier Number: CT-RG-100
  • 76197-168
    Disposable Trough Liner
    Disposable Trough Liner
    PP
    100 ml
    Log in for availability
    Log in for availability
    Case of 1,600
    Case of 1,600 - $946.47
    $946.47
    New Customer Promotional Price
    Case of 1,600
       $946.47
    Supplier Number: CT-L-100