Bio-Safe NA Complete Counting Cocktail, RPI
Catalog # 61000-506
Supplier: RPI
Small Business Enterprise
CAS Number:
Specifications
- Description:Bio-Safe NA Complete Counting Cocktail
- Size:4 L
- Cat. no.:61000-506
- Supplier No.:111198
Specifications
About this item
Bio-Safe NA provides the highest efficiency for counting organic samples, dry filters and samples treated with tissue solubilizers.
- Bluish tint liquid
- Odorless
Effective and economical cocktail for counting dry wipe tests in radiation safety procedures. Compatible with all plastic scintillation vials.
Caution: Research or further manufacturing use only, not for food or drug use.