Fixation Buffer, Biolegend
Fixation Buffer, 100 mL
Catalog # 420801-BL
Supplier: Biolegend
Supplier #: 420801
Jump to:
Product Details & Documents
Fixation Buffer is useful for intracellular staining procedures like preparation of cells for staining intracellular cytokines or other proteins and to fix cells prior to permeabilization using permeabilization wash buffer.
Specifications
Specifications
- Description:Fixation Buffer
- Size:100 mL
- Application:For Intracellular Staining Procedures and to Fix Cells
- Cat. No.:420801-BL
- Storage temperature:2 – 8 °C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



