Substrate for Western Blotting, Trident Plus, GeneTex
Catalog # 10793-298
Supplier: Genetex
CAS Number:
Specifications
- Description:Trident plus Western HRP Substrate
- Size:500 mL
- Cat. no.:10793-298
Specifications
About this item
Trident Plus Western HRP substrate is a luminol-based, chemiluminescent substrate for the detection of HRP-conjugated antibody on immunoblots at picogram level. It produces an excellent signal that can be easily detected by chemiluminescent imaging methods including X-ray film and chemiluminescence imaging system.
- Liquid form
- Used in western blot
- Store at 4 °C
Contains Trident Plus Western HRP solution A (250 mL) and Trident Plus Western HRP substrate solution B (250 mL).
Caution: For In vitro laboratory use only. Not for any clinical, therapeutic or diagnostic use in humans or animals. Not for animal or human consumption.